missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Chloroethyl)morpholine hydrochloride, 99%

Product Code. 11377585
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377585 100 g 100g
11387585 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11377585 Supplier Thermo Scientific Alfa Aesar Supplier No. L05771.22
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 3647-69-6 | C6H13Cl2NO | 186.076 g/mol

4-(2-Chloroethyl)morpholine hydrochloride (MOC) is used as intermediate for the synthesis of pharmaceuticals (e.g. floredil, morinamide, nimorazole and pholcodine). It is also used as Agrochemical Intermediates, dyestuff Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(2-Chloroethyl)morpholine hydrochloride (MOC) is used as intermediate for the synthesis of pharmaceuticals (e.g. floredil, morinamide, nimorazole and pholcodine). It is also used as Agrochemical Intermediates, dyestuff Intermediates.

Solubility
Soluble in water, and methanol.

Notes
Hygroscopic. Do not store together with acids. Store under dry inert gas. Protect from humidity and water. Incompatible materials are acids, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3647-69-6
Molecular Formula C6H13Cl2NO
Molecular Weight (g/mol) 186.076
MDL Number MFCD00012797
InChI Key NBJHDLKSWUDGJG-UHFFFAOYSA-N
Synonym 4-2-chloroethyl morpholine hydrochloride, n-2-chloroethyl morpholine hydrochloride, 2-morpholinoethylchloride hydrochloride, morpholine, 4-2-chloroethyl-, hydrochloride, 4-2-chloroethyl morpholine hcl, chloromorpholinoethane hydrochloride, 2-chloroethyl morpholine hydrochloride, 2-morpholinoethyl chloride hydrochloride, 4-2-chloroethyl-morpholine hydrochloride, 4-2-chloroethyl morpholinium chloride
PubChem CID 77210
IUPAC Name 4-(2-chloroethyl)morpholine;hydrochloride
SMILES C1COCCN1CCCl.Cl

Specifications

Melting Point 184°C to 186°C
Quantity 100 g
UN Number UN2923
Beilstein 3684083
Sensitivity Hygroscopic
Solubility Information Soluble in water,and methanol.
Formula Weight 186.08
Percent Purity 99%
Chemical Name or Material 4-(2-Chloroethyl)morpholine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.