missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Cyano-3-ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid pinacol ester, Thermo Scientific™

Product Code. 15419185
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15419185 1 g 1g
15429185 5 g 5g
15439185 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15419185 Supplier Thermo Scientific Alfa Aesar Supplier No. H51130.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1218790-51-2
Molecular Formula C18H22BNO4
Molecular Weight (g/mol) 327.187
MDL Number MFCD11113044
InChI Key DNALFJBPPFISJQ-SDNWHVSQSA-N
Synonym ethyl 2e-2-cyano-3-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl prop-2-enoate, 4-2-cyano-3-ethoxy-3-oxo-1-propen-1-yl benzeneboronic acid pinacol ester, 2-cyano-3-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-acrylic acid ethyl ester, amtb130, 2-cyano-3-4-4,4,5,5-tetramethyl-1,3,2 dioxa-borolan-2-yl-phenyl-acrylic acid ethyl ester, e-4-2-cyano-2-ethoxycarbonylvinyl phenyl boronic acid pinacol ester, e-ethyl 2-cyano-3-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl acrylate, ethyl 2e-2-cyano-3-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl prop-2-enoate
PubChem CID 46739750
IUPAC Name ethyl (E)-2-cyano-3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]prop-2-enoate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C=C(C#N)C(=O)OCC

Specifications

Quantity 1 g
Formula Weight 327.18
Chemical Name or Material 4-(2-Cyano-3-ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.