missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-[2-(Dimethylamino)ethoxy]benzaldehyde, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | 4-[2-(Dimethylamino)ethoxy]benzaldehyde |
| CAS | 15182-92-0 |
| Molecular Formula | C11H15NO2 |
| Quantity | 250 mg |
| Synonym | p-[2-(Dimethylamino)ethoxy]benzaldehyde |
| InChI Formula | InChI=1S/C11H15NO2/c1-12(2)7-8-14-11-5-3-10(9-13)4-6-11/h3-6,9H,7-8H2,1-2H3 |
| SMILES | CN(C)CCOc1ccc(C=O)cc1 |
| IUPAC Name | 4-[2-(dimethylamino)ethoxy]benzaldehyde |
| Molecular Weight (g/mol) | 193.24 |
| Formula Weight | 193.1103 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction