missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methoxyethyl)aniline, TRC

Product Code. 30478175
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478175 5 g 5g
30483067 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30478175 Supplier Toronto Research Chemicals Supplier No. TRCM2272005G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84803-56-5
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
Synonym 4-(2-Methoxyethyl)benzenamine, p-2-Methoxyethyl-Aniline, 1-Methoxy-2-(4-aminophenyl)ethane, 4-(2-Methoxyethyl)phenylamine, p-(2-Methoxyethyl)aniline
IUPAC Name 4-(2-methoxyethyl)aniline
SMILES COCCc1ccc(N)cc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C9H13NO/c1-11-7-6-8-2-4-9(10)5-3-8/h2-5H,6-7,10H2,1H3
Formula Weight 151.0997
Chemical Name or Material 4-(2-Methoxyethyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.