missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methoxyethylaminocarbonyl)benzeneboronic acid pinacol ester, 97%

Product Code. 11377539
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377539 5 g 5g
11367539 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377539 Supplier Thermo Scientific Alfa Aesar Supplier No. H27945.06
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1073353-60-2 | C16H24BNO4 | 305.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073353-60-2
Molecular Formula C16H24BNO4
Molecular Weight (g/mol) 305.18
MDL Number MFCD09266188
InChI Key XNDAIQVEKVZONM-UHFFFAOYSA-N
Synonym n-2-methoxyethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 4-2-methoxyethylcarbamoyl phenylboronic acid, pinacol ester, 4-2-methoxyethylamine-1-carbonyl phenyl boronic acid pinacol ester, amtb042, 4-2-methoxyethyl carbamoyl phenylboronic acid pinacol ester, 4-2-methoxyethylaminecarbonyl benzeneboronic acid pinacol ester, n-2-methoxyethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide
PubChem CID 46739044
IUPAC Name N-(2-methoxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
SMILES COCCNC(=O)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Tekniske data

Melting Point 73°C to 76°C
Quantity 5 g
Formula Weight 305.18
Percent Purity 97%
Chemical Name or Material 4-(2-Methoxyethylaminocarbonyl)benzeneboronic acid pinacol ester

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.