missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methylphenyl)-3-thiosemicarbazide, 98%, Thermo Scientific™

Product Code. 11357396
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357396 25 g 25g
11347396 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357396 Supplier Thermo Scientific Alfa Aesar Supplier No. L09382.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 614-10-8
Molecular Formula C8H11N3S
Molecular Weight (g/mol) 181.257
MDL Number MFCD00041277
InChI Key QCMQZMITIOLXFZ-UHFFFAOYSA-N
Synonym 4-2-methylphenyl-3-thiosemicarbazide, n-o-tolyl hydrazinecarbothioamide, 3-amino-1-2-methylphenyl thiourea, n-2-methylphenyl hydrazinecarbothioamide, hydrazino 2-methylphenyl amino methane-1-thione, acmc-1bcwm, wln: zmyus&mr b1, 4-o-tolyl thiosemicarbazide, 1-amino-3-2-methylphenyl thiourea, hydrazinecarbothioamide,n-2-methylphenyl
PubChem CID 737133
IUPAC Name 1-amino-3-(2-methylphenyl)thiourea
SMILES CC1=CC=CC=C1NC(=S)NN

Specifications

Melting Point 145°C to 148°C
Quantity 25 g
UN Number UN2811
Beilstein 880437
Formula Weight 181.26
Percent Purity 98%
Chemical Name or Material 4-(2-Methylphenyl)-3-thiosemicarbazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.