missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Morpholin-4-ylethoxy)benzonitrile, 95%, Thermo Scientific™

Product Code. 10351523
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10351523 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10351523 Supplier Thermo Scientific Maybridge Supplier No. CC42116DA
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 34334-04-8
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C13H16N2O2
MDL Number MFCD07772850
Quantity 1 g
Synonym 4-2-morpholin-4-ylethoxy benzonitrile, 4-2-morpholinoethoxy benzonitrile, 4-2-morpholin-4-yl ethoxy benzonitrile, 4-2-4-morpholinyl ethoxy benzonitrile, 4-2-morpholin-4-yl-ethoxy-benzonitrile, benzonitrile,4-2-4-morpholinyl ethoxy, 4-2-morpholin-4-ylethoxy benzenecarbonitrile, benzonitrile, 4-2-4-morpholinyl ethoxy
InChI Key KQTJRPZKCNRDQC-UHFFFAOYSA-N
SMILES C1COCCN1CCOC2=CC=C(C=C2)C#N
IUPAC Name 4-(2-morpholin-4-ylethoxy)benzonitrile
Molecular Weight (g/mol) 232.283
PubChem CID 2994074
Formula Weight 232.28
Percent Purity 95%
Chemical Name or Material 4-(2-Morpholin-4-ylethoxy)benzonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.