missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(2-Nitrophenylsulfonyl)morpholine, 97%

Brand:  Alfa Aesar™ H60520.03

Code : NEW

Additional Details : CAS Number : 99069-26-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15416998

  • 448.12 € / 1g


Estimated Shipment date
from Supplier 30-07-2018
Add to basket

Specifications

Chemical Identifiers

4-(2-Nitrophenylsulfonyl)morpholine
99069-26-8
MFCD00514601

Specifications

4-(2-Nitrophenylsulfonyl)morpholine
99069-26-8
97%
MFCD00514601
NXJKHLDWXHCTMN-UHFFFAOYSA-N
272.275
272.28
Solid
C10H12N2O5S
1g
C1COCCN1S(=O)(=O)C2=CC=CC=C2[N+](=O)[O-]
796972
Videos
SDS