missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Pyridylazo)resorcinol, TRC

Product Code. 30424742
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424742 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30424742 Supplier Toronto Research Chemicals Supplier No. TRCP991998100MG
 Restricted Item
Estimated Shipment: 30-11-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material 4-(2-Pyridylazo)resorcinol
CAS 1141-59-9
Molecular Formula C11H9N3O2
Quantity 100 mg
InChI Formula InChI=1S/C11H9N3O2/c15-8-4-5-9(10(16)7-8)13-14-11-3-1-2-6-12-11/h1-7,15-16H
SMILES Oc1ccc(N=Nc2ccccn2)c(O)c1
IUPAC Name 4-[(E)-pyridin-2-yldiazenyl]benzene-1,3-diol
Molecular Weight (g/mol) 215.21
Formula Weight 215.0695
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.