missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Pyrrolidinyl)pyridine, 96%

Product Code. 15463357
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15463357 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15463357 Supplier Thermo Scientific Alfa Aesar Supplier No. H26051.MD
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 128562-25-4 | C9H12N2 | 148.209 g/mol

4-(2-Pyrrolidinyl)pyridine can be used in aldol catalysis, in the preparation of azole compounds as 11β-HSD1 inhibitors as well as in the asymetric synthesis of pyrazolodihydropyrimidinylpyrrolidin amide derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(2-Pyrrolidinyl)pyridine can be used in aldol catalysis, in the preparation of azole compounds as 11β-HSD1 inhibitors as well as in the asymetric synthesis of pyrazolodihydropyrimidinylpyrrolidin amide derivatives.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store in a cool, dark and well-ventilated place. Store away from incompatible materials such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128562-25-4
Molecular Formula C9H12N2
Molecular Weight (g/mol) 148.209
MDL Number MFCD01862536
InChI Key GDGNPIOGJLCICG-UHFFFAOYSA-N
Synonym 4-pyrrolidin-2-yl pyridine, 4-2-pyrrolidinyl pyridine, 4-pyrrolidin-2-yl-pyridine, 2-4-pyridyl pyrrolidine, pyridine, 4-2-pyrrolidinyl, 2-4-pyridinyl pyrrolidine, +/--4-pyrrolidin-2-yl pyridine, 4-2-pyridyl pyrrolidine, 2-pyridin-4-yl pyrrolidine
PubChem CID 2771664
IUPAC Name 4-pyrrolidin-2-ylpyridine
SMILES C1CC(NC1)C2=CC=NC=C2

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 148.21
Percent Purity 96%
Chemical Name or Material 4-(2-Pyrrolidinyl)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.