missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Tetrahydropyranyloxy)phenylmagnesium bromide, 0.5M in 2-MeTHF

Product Code. 15410786
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15410786 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15410786 Supplier Thermo Scientific Alfa Aesar Supplier No. H54115.AE
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 36637-44-2 | C11H13BrMgO2 | 281.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 36637-44-2
Molecular Formula C11H13BrMgO2
MDL Number MFCD03427254
Quantity 100 mL
UN Number UN2924
Beilstein 3547417
Sensitivity Air and moisture sensitive
Synonym 4-2-tetrahydro-2h-pyranoxy phenylmagnesium bromide, ohpuqubykzjoly-uhfffaoysa-m, 2-4-bromomagnesio phenoxy oxane, p-2-tetrahydropyranyloxy phenyl magnesium bromide, 4-2-tetrahydro-3h-pyranoxy phenylmagnesium bromide, 4-tetrahydropyran-2-yloxy phenyl magnesium bromide, 4-2-tetrahydro-3-h-pyranoxy phenylmagnesium bromide, 4-tetrahydro-2h-pyran-2-yloxy phenyl magnesium bromide, 4-2-tetrahydro-2h-pyranoxy phenylmagnesium bromide solution, 4-2-tetrahydropyranyloxy phenylmagnesium bromide, 0.5m in 2-methf, grignard reagent
InChI Key OHPUQUBYKZJOLY-UHFFFAOYNA-M
SMILES Br[Mg]C1=CC=C(OC2CCCCO2)C=C1
IUPAC Name magnesium;2-(phenoxy)oxane;bromide
Molecular Weight (g/mol) 281.43
PubChem CID 14519569
Formula Weight 281.43
Concentration or Composition (by Analyte or Components) 0.5M in 2-MeTHF
Chemical Name or Material 4-(2-Tetrahydropyranyloxy)phenylmagnesium bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.