missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Thienyl)benzenesulfonyl chloride, 96%

Product Code. 15477337
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15477337 250 mg 250mg
15457337 1 g 1g
15467337 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15477337 Supplier Thermo Scientific Alfa Aesar Supplier No. H34399.MD
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 209592-91-6 | C10H7ClO2S2 | 258.734 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 209592-91-6
Molecular Formula C10H7ClO2S2
Molecular Weight (g/mol) 258.734
MDL Number MFCD11655569
InChI Key IRDZCMUQNGEQEP-UHFFFAOYSA-N
Synonym 4-2-thienyl benzenesulfonyl chloride, benzenesulfonyl chloride, 4-2-thienyl, 4-thiophen-2-yl benzene-1-sulfonyl chloride, 4-thiophen-2-yl benzenesulfonyl chloride
PubChem CID 18387759
IUPAC Name 4-thiophen-2-ylbenzenesulfonyl chloride
SMILES C1=CSC(=C1)C2=CC=C(C=C2)S(=O)(=O)Cl

Specifications

Melting Point 107°C to 108°C
Quantity 250 mg
UN Number UN3261
Formula Weight 258.75
Percent Purity 96%
Chemical Name or Material 4-(2-Thienyl)benzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.