missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Thienyl)butyric acid, 98%

Product Code. 11391947
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11391947 1 g 1g
11301957 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11391947 Supplier Thermo Scientific Alfa Aesar Supplier No. L07563.03
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 4653-11-6 | C8H9O2S | 169.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4653-11-6
MDL Number MFCD00005463
ChEBI CHEBI:40114

Specifications

Density 1.169
Boiling Point 122°C (0.3 mmHg)
Flash Point >110°C (230°F)
Molecular Formula C8H9O2S
Refractive Index 1.532
Quantity 1 g
Beilstein 127982
Synonym 4-2-thienyl butyric acid, 4-thiophen-2-yl butanoic acid, 2-thiophenebutanoic acid, 2-thiophenebutyric acid, 4-2-thienyl butanoic acid, 4-thiophen-2-yl-butyric acid, 4-thiophen-2-yl-butyricacid, gamma-alpha-thienyl butyricacid, .gamma.-.alpha.-thienyl butyric acid, 4tb
InChI Key VYTXLSQVYGNWLV-UHFFFAOYSA-M
SMILES C1=CSC(=C1)CCCC(=O)O
IUPAC Name 4-thiophen-2-ylbutanoic acid
Molecular Weight (g/mol) 169.22
PubChem CID 78386
Formula Weight 170.23
Percent Purity 98%
Chemical Name or Material 4-(2-Thienyl)butyric acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.