missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ 4 - 20 mA Current Loop Interface

Product Code. 17110125 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP1400eco
HPP2200eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP110eco
IPP1400eco
IPP2200eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17110125

Brand: Memmert™ IPP110ecoV3

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

4-20mA current loop interface temperature controller

  • Temperature controller
  • Actual value; -10 to +80 °C = 4 -20 mA

Specifications

For Use With (Equipment) IPP110eco
Type Temperature Controller
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.