missing translation for 'onlineSavingsMsg'
Learn More

4-(2H-Tetrazol-5-yl)benzeneboronic acid pinacol ester, 95%

Product Code. 15454896
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15454896 250 mg 250mg
15444896 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15454896 Supplier Thermo Scientific Alfa Aesar Supplier No. H59222.MD
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 775351-40-1 | C13H17BN4O2 | 272.115 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 775351-40-1
Molecular Formula C13H17BN4O2
Molecular Weight (g/mol) 272.115
MDL Number MFCD12761127
InChI Key ONAVJWUKLJDJAI-UHFFFAOYSA-N
Synonym 5-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-2h-tetrazole, 4-1h-tetrazol-5-yl benzene-1-boronic acid pinacol ester, 4-2h-tetrazol-5-yl benzeneboronic acid pinacol ester, 5-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-1,2,3,4-tetrazole, 4-2h-tetrazol-5-yl phenylboronic acid pinacol ester, 5-4-phenylboronic acid pinacol ester-2h-tetrazole, 4-1h-tetrazol-5-yl phenyl boronic acid pinacol ester, 5-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-tetrazole, 5-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-1,2,3,4-tetrazole
PubChem CID 45785702
IUPAC Name 5-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-2H-tetrazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C3=NNN=N3

Specifications

Quantity 250 mg
Formula Weight 272.11
Percent Purity 95%
Chemical Name or Material 4-(2H-Tetrazol-5-yl)benzeneboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.