missing translation for 'onlineSavingsMsg'
Learn More

4-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine, 90%, Thermo Scientific™

Product Code. 10586524
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10586524 5 g Amber Glass Bottle 5g
10096342 250 mg Amber Glass Bottle 250mg
10216752 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10586524 Supplier Thermo Scientific Maybridge Supplier No. CC33839DE
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 852227-95-3
Molecular Formula C16H24BNO3
Molecular Weight (g/mol) 289.18
MDL Number MFCD03412097
InChI Key NCJDKFFODGZRRL-UHFFFAOYSA-N
Synonym 4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine, 3-morpholinophenylboronic acid pinacol ester, 3-morpholino phenylboronic acid pinacol ester, 3-4-morpholino phenylboronic acid pinacol ester, 4-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine, pubchem19450, ksc496k1f, 3-morpholinophenylboronic acidpinacolester, 3-morpholinophenylboronic acid pinacol
PubChem CID 4192663
IUPAC Name 4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine
SMILES CC1(C)OB(OC1(C)C)C1=CC(=CC=C1)N1CCOCC1

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 289.18
Percent Purity 90%
Chemical Name or Material 4-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.