missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Bromothien-2-yl)benzoic acid, 97%, Thermo Scientific™

Product Code. 10113044
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10113044 1 g Amber Glass Bottle 1g
10622505 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10113044 Supplier Thermo Scientific Maybridge Supplier No. CC67801DA
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 930111-09-4
Molecular Formula C11H7BrO2S
Molecular Weight (g/mol) 283.14
MDL Number MFCD09879966
InChI Key WAOAWGITQLHUFM-UHFFFAOYSA-N
Synonym 4-3-bromothien-2-yl benzoic acid, 4-3-bromothiophen-2-yl benzoic acid
PubChem CID 24229749
IUPAC Name 4-(3-bromothiophen-2-yl)benzoic acid
SMILES OC(=O)C1=CC=C(C=C1)C1=C(Br)C=CS1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 283.14
Percent Purity 97%
Chemical Name or Material 4-(3-Bromothien-2-yl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.