missing translation for 'onlineSavingsMsg'
Learn More

4-[3-Chloro-4-(3-fluorobenzyloxy)phenylamino]-6-iodoquinazoline, 97%

Product Code. 15480926
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15480926 250 mg 250mg
15470926 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15480926 Supplier Thermo Scientific Alfa Aesar Supplier No. H54229.MD
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 231278-20-9 | C21H14ClFIN3O | 505.72 g/mol

4-[3-Chloro-4-(3-fluorobenzyloxy)phenylamino]-6-iodoquinazoline is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-[3-Chloro-4-(3-fluorobenzyloxy)phenylamino]-6-iodoquinazoline is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 231278-20-9
Molecular Formula C21H14ClFIN3O
Molecular Weight (g/mol) 505.72
MDL Number MFCD09998827
InChI Key UHFPFDMMKYQMLC-UHFFFAOYSA-N
Synonym n-3-chloro-4-3-fluorobenzyl oxy phenyl-6-iodoquinazolin-4-amine, n-3-chloro-4-3-fluorobenzyloxy phenyl-6-iodoquinazolin-4-amine, n-3-chloro-4-3-fluorobenzyloxy-phenyl-6-iodoquinazolin-4-amine, n-3-chloro-4-3-fluorophenyl methoxy phenyl-6-iodoquinazolin-4-amine, 6-iodo-4-3-chloro-4-3-fluorobenzyloxy anilino quinazoline, 4-3-chloro-4-3-fluorobenzyloxy phenylamino-6-iodoquinazoline, n-3-chloro-4-3-fluorobenzyl oxy phenyl-6-iodo-4-quinazolinamine, 4-quinazolinamine, n-3-chloro-4-3-fluorophenyl methoxy phenyl-6-iodo
PubChem CID 10174519
IUPAC Name N-[3-chloro-4-[(3-fluorophenyl)methoxy]phenyl]-6-iodoquinazolin-4-amine
SMILES FC1=CC=CC(COC2=C(Cl)C=C(NC3=C4C=C(I)C=CC4=NC=N3)C=C2)=C1

Specifications

Quantity 250 mg
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 505.72
Percent Purity 97%
Chemical Name or Material 4-[3-Chloro-4-(3-fluorobenzyloxy)phenylamino]-6-iodoquinazoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.