missing translation for 'onlineSavingsMsg'
Learn More

4-((3-Chloro-4-fluorophenyl)amino)-7-methoxyquinazolin-6-ol, 98%, BLD Pharm™

Product Code. 30797286
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
250mg
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30797286 1 g 1g
30796330 100 g 100g
30785268 5 g 5g
30796375 25 g 25g
30785468 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30797286 Supplier BLD Pharm Supplier No. BD1461201g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 184475-71-6
Molecular Formula C15H11ClFN3O2
MDL Number MFCD09908046
Quantity 1 g
SMILES OC1=CC2=C(NC3=CC=C(F)C(Cl)=C3)N=CN=C2C=C1OC
Molecular Weight (g/mol) 319.72
Percent Purity 98%
Chemical Name or Material 4-((3-Chloro-4-fluorophenyl)amino)-7-methoxyquinazolin-6-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.