missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Methyl-1,2,4-oxadiazol-5-yl)benzoic acid, 95%, Thermo Scientific™

Product Code. 10654595
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10654595 10 g 10g
10545384 250 mg 250mg
10184054 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10654595 Supplier Thermo Scientific Maybridge Supplier No. CC50901EA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 851048-56-1
Molecular Formula C10H8N2O3
Molecular Weight (g/mol) 204.19
MDL Number MFCD08741427
InChI Key KXXLPIJOPGBFTA-UHFFFAOYSA-N
Synonym 4-3-methyl-1,2,4-oxadiazol-5-yl benzoic acid, benzoicacid, 4-3-methyl-1,2,4-oxadiazol-5-yl, benzoic acid, 4-3-methyl-1,2,4-oxadiazol-5-yl, methyloxadiazolylbenzoicacid, 4-3-methyl-1,2,4 oxadiazol-5-yl-benzoic acid, 4-3-methyl-1,2,4-oxadiazol-5-yl-benzoic acid
PubChem CID 22110111
IUPAC Name 4-(3-methyl-1,2,4-oxadiazol-5-yl)benzoic acid
SMILES CC1=NOC(=N1)C1=CC=C(C=C1)C(O)=O

Specifications

Assay Percent Range 95%
Quantity 10 g
Formula Weight 204.18
Chemical Name or Material 4-(3-Methyl-1, 2, 4-oxadiazol-5-yl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.