missing translation for 'onlineSavingsMsg'
Learn More

4-[3-(Trifluoromethyl)diazirin-3-yl]benzoic Acid N-Hydroxysuccinimide Ester, TRC

Product Code. 30457214
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457214 5 mg 5mg
30452242 2.5 mg 2.5mg
30451674 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30457214 Supplier Toronto Research Chemicals Supplier No. TRCT7912505MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87736-89-8
Molecular Formula C13 H8 F3 N3 O4
Molecular Weight (g/mol) 327.22
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 4-[3-(trifluoromethyl)diazirin-3-yl]benzoate
SMILES FC(F)(F)C1(N=N1)c2ccc(cc2)C(=O)ON3C(=O)CCC3=O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C13H8F3N3O4/c14-13(15,16)12(17-18-12)8-3-1-7(2-4-8)11(22)23-19-9(20)5-6-10(19)21/h1-4H,5-6H2
Formula Weight 327.0467
Chemical Name or Material 4-[3-(Trifluoromethyl)diazirin-3-yl]benzoic Acid N-Hydroxysuccinimide ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.