missing translation for 'onlineSavingsMsg'
Learn More

4-[(3S)-3-[[(1R)-1-(1-Naphthalenyl)ethyl]amino]-1-pyrrolidinyl]benzeneacetic Acid, TRC

Product Code. 30476313
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30476313 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30476313 Supplier Toronto Research Chemicals Supplier No. TRCN345665100MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-((3S)-3-(((1R)-1-(1-Naphthalenyl)ethyl)amino)-1-pyrrolidinyl)benzeneacetic Acid
CAS 870964-67-3
Molecular Formula C24 H26 N2 O2
Quantity 100 mg
Synonym Benzeneacetic acid','4-[(3S)-3-[[(1R)-1-(1-naphthalenyl)ethyl]amino]-1-pyrrolidinyl]- (9CI','ACI), 4-[(3S)-3-[[(1R)-1-(1-Naphthalenyl)ethyl]amino]-1-pyrrolidinyl]benzeneacetic acid (ACI), Evocalcet, Orkedia
InChI Formula InChI=1S/C24H26N2O2/c1-17(22-8-4-6-19-5-2-3-7-23(19)22)25-20-13-14-26(16-20)21-11-9-18(10-12-21)15-24(27)28/h2-12,17,20,25H,13-16H2,1H3,(H,27,28)/t17-,20+/m1/s1
SMILES C[C@@H](N[C@H]1CCN(C1)c2ccc(CC(=O)O)cc2)c3cccc4ccccc34
IUPAC Name 2-[4-[(3S)-3-[[(1R)-1-naphthalen-1-ylethyl]amino]pyrrolidin-1-yl]phenyl]acetic acid
Molecular Weight (g/mol) 374.475
Formula Weight 374.199
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.