missing translation for 'onlineSavingsMsg'
Learn More

4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyran, Thermo Scientific™

Product Code. 10371650
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10371650 1 g Amber Glass Bottle 1g
10505660 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10371650 Supplier Thermo Scientific Maybridge Supplier No. MO08545DA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 287944-16-5
Molecular Formula C11H19BO3
Molecular Weight (g/mol) 210.08
MDL Number MFCD11052631
InChI Key DOSGEBYQRMBTGS-UHFFFAOYSA-N
Synonym 3,6-dihydro-2h-pyran-4-boronic acid pinacol ester, 2-3,6-dihydro-2h-pyran-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-pyran, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyran, 2h-pyran, 3,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyran, 3,6-dihydro-2h-pyran-4-boronic acid, pinacol ester, 3,6-dihydro-2h-pyran-4-boronicacidpinacolester, 3,6-dihydro-2h-pyran-4-boronic acid pinacolester, 3,6 dihydro-2h-pyran-4-boronic acid pinacol ester
PubChem CID 11218053
IUPAC Name 2-(3,6-dihydro-2H-pyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CCOCC1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 210.08
Percent Purity 95%
Chemical Name or Material 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.