missing translation for 'onlineSavingsMsg'
Learn More

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid, 97%

Product Code. 10457592 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10457592 1 g Glass bottle 1g
10394782 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10457592 Supplier Thermo Scientific Acros Supplier No. 366390010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 180516-87-4 | C13H17BO4 | 248.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 180516-87-4
Molecular Formula C13H17BO4
Molecular Weight (g/mol) 248.09
MDL Number MFCD01863710
InChI Key IYDKBQIEOBXLTP-UHFFFAOYSA-N
Synonym 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 4-carboxyphenylboronic acid pinacol ester, 4-carboxylphenylboronic acid pinacol ester, 4-carboxyphenylboronic acid, pinacol ester, 4-carboxyphenylboronicacidpinacolester, 4-carboxybenzeneboronic acid pinacol ester, 4-carboxyphenylboronic acid pinacolate, 4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 4-carboxyphenylboronic acid,pinacol ester, 4-carboxyphenyl boronic acid, pinacol ester
PubChem CID 2734621
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C(O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 227.0°C to 232.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble.
Formula Weight 248.09
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.