missing translation for 'onlineSavingsMsg'
Learn More

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 97%

Product Code. 10552992 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10552992 5 g Glass bottle 5g
10202382 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10552992 Supplier Thermo Scientific Acros Supplier No. 367510050
 Restricted Item
In Stock
Læg i indkøbskurv
Læg i indkøbskurv
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

CAS: 181219-01-2 | C11H16BNO2 | 205.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 181219-01-2
Molecular Formula C11H16BNO2
Molecular Weight (g/mol) 205.06
MDL Number MFCD01319051
InChI Key NLTIETZTDSJANS-UHFFFAOYSA-N
Synonym 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 4-pyridineboronic acid pinacol ester, pyridine-4-boronic acid pinacol ester, 4-pyridylboronic acid pinacol ester, pyridine-4-boronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine-4-boronic pinacolate, pyridine-4-boronic acid pinacol cyclic ester, 2-4-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, pyridine-4-boronic pinacol ester
PubChem CID 2734648
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CC1(C)OB(OC1(C)C)C1=CC=NC=C1

Tekniske data

CAS Min % 96.0
CAS Max % 100.0
Melting Point 153.0°C to 155.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 205.06
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Vis mere Vis mindre

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.