missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)tetrahydropyran, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Specifications
Specifications
| Assay Percent Range | 97% |
| Quantity | 250 mg |
| Formula Weight | 212.09 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)tetrahydropyran |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction