missing translation for 'onlineSavingsMsg'
Learn More

4,4,4-Trifluoro-2-methylbutyric acid, 95%, Thermo Scientific™

Product Code. 11323766
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11323766

Brand: Thermo Scientific Alfa Aesar L16813.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4,4,4-Trifluoro-2-methylbutyric acid, is used as pharma intermediates

Solubility
Soluble in water and ethanol, and insoluble in ether

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99783-23-0
Molecular Formula C5H7F3O2
Molecular Weight (g/mol) 156.104
MDL Number MFCD00190640
InChI Key OBUKEMPWKGDRJV-UHFFFAOYSA-N
Synonym 2-methyl-4,4,4-trifluorobutyric acid, 4,4,4-trifluoro-2-methyl-butanoic acid, 2-methyl-4,4,4-trifluorobutanoic acid, 4,4,4-trifluoro-2-methylbutyric acid, 2-methyl-4,4,4-trifluorobutanoic acid, butanoic acid, 4,4,4-trifluoro-2-methyl, 4,4,4-tris fluoranyl-2-methyl-butanoic acid
PubChem CID 2775536
IUPAC Name 4,4,4-trifluoro-2-methylbutanoic acid
SMILES CC(CC(F)(F)F)C(=O)O

Specifications

Boiling Point 172°C to 175°C
Refractive Index 1.358
Quantity 1 g
UN Number UN3265
Beilstein 6917955
Solubility Information Soluble in water and ethanol,and insoluble in ether
Formula Weight 156.11
Percent Purity 95%
Chemical Name or Material 4,4,4-Trifluoro-2-methylbutyric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.