missing translation for 'onlineSavingsMsg'
Learn More

4,4,4-Trifluoro-3-hydroxy-3-(trifluoromethyl)butyric acid, 97%

Product Code. 11387466
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387466 5 g 5g
11377466 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387466 Supplier Thermo Scientific Alfa Aesar Supplier No. L09516.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1547-36-0 | C5H4F6O3 | 226.07 g/mol

4,4,4-Trifluoro-3-hydroxy-3-(trifluoromethyl)butyric acid is a beta-hydroxy acid undergoes cyclization reaction to prepare beta-lactone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4,4-Trifluoro-3-hydroxy-3-(trifluoromethyl)butyric acid is a beta-hydroxy acid undergoes cyclization reaction to prepare beta-lactone.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1547-36-0
Molecular Formula C5H4F6O3
Molecular Weight (g/mol) 226.07
MDL Number MFCD00040910
InChI Key NOHJBOWARMTILE-UHFFFAOYSA-N
PubChem CID 15244
IUPAC Name 4,4,4-trifluoro-3-hydroxy-3-(trifluoromethyl)butanoic acid
SMILES OC(=O)CC(O)(C(F)(F)F)C(F)(F)F

Specifications

Melting Point 85°C to 89°C
Quantity 5 g
UN Number UN3261
Beilstein 1911706
Synonym 3,3-bis trifluoromethyl-3-hydroxypropionic acid, 4,4,4-trifluoro-3-hydroxy-3-trifluoromethyl butanoic acid, butyric acid, 3-hydroxy-4,4,4-trifluoro-3-trifluoromethyl, 3-hydroxy-4,4,4-trifluoro-3-trifluoromethyl butyric acid, 4,4,4-trifluoro-3-hydroxy-3-trifluoromethyl butyric acid, 4,4,4-trifluoro-3-hydroxy-3-trifluoromethylbutyric acid, butanoic acid, 4,4,4-trifluoro-3-hydroxy-3-trifluoromethyl, butanoic acid,4,4,4-trifluoro-3-hydroxy-3-trifluoromethyl, acmc-20anh2, 3,3-bis trifluoromethyl-3-hydroxypropanoic acid
Solubility Information Soluble in water.
Formula Weight 226.08
Percent Purity 97%
Chemical Name or Material 4,4,4-Trifluoro-3-hydroxy-3-(trifluoromethyl)butyric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.