missing translation for 'onlineSavingsMsg'
Learn More

4,4,4-Trifluorobutylamine, 97%

Product Code. 11975181 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11975181 5 g Glass bottle 5g
11965181 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11975181 Supplier Thermo Scientific Acros Supplier No. 432590050
 Restricted Item
Estimated Shipment: 12-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 819-46-5 | C4H8F3N | 127.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 819-46-5
Molecular Formula C4H8F3N
Molecular Weight (g/mol) 127.11
InChI Key LAXWLCVPJLBABV-UHFFFAOYSA-N
PubChem CID 136645
IUPAC Name 4,4,4-trifluorobutan-1-amine
SMILES C(CC(F)(F)F)CN

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Boiling Point 90°C to 94°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Synonym 4,4,4-trifluorobutylamine, 4,4,4-trifluorobutyl-1-amine, 1-butanamine,4,4,4-trifluoro, cf3ch2ch2ch2nh2, 1-butanamine, 4,4,4-trifluoro, 4,4,4-trifluoro-butylamine, 4,4,4-trifluorobutyl-amine, 4,4,4-trifluoro-1-butanamine, 1,1,1-trifluorobutane-4-amine, 1-amino-4,4,4-trifluorobutane
Formula Weight 127.11
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4, 4, 4-Trifluorobutylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4,4,4-Trifluorobutylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.