missing translation for 'onlineSavingsMsg'
Learn More

4,4,4-Trifluorocrotononitrile, 96%

Product Code. 15469049
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15469049 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15469049 Supplier Thermo Scientific Alfa Aesar Supplier No. L16870.03
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 406-86-0 | C4H2F3N | 121.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 406-86-0
Boiling Point 84°C to 85°C
Molecular Formula C4H2F3N
MDL Number MFCD00190645
Quantity 1 g
UN Number UN3276
Beilstein 6381034
Synonym 4,4,4-trifluorocrotononitrile, 4,4,4-trifluorocrotonitrile, 2e-4,4,4-trifluorobut-2-enenitrile, 3-trifluoromethylacrylonitrile, 4,4,4-trifluorobut-2-enenitrile, 4,4,4-trifluoro-2-butenenitrile, trifluorocrotononitrile, ka, a`\d`dadadtarjfppp, e-4,4,4-trifluoro-2-butenenitrile, e-4,4,4-trifluorobut-2-enenitrile
InChI Key LHWSEFCIRYVTLZ-UHFFFAOYSA-N
SMILES FC(F)(F)C=CC#N
IUPAC Name (E)-4,4,4-trifluorobut-2-enenitrile
Molecular Weight (g/mol) 121.06
PubChem CID 5708782
Formula Weight 121.06
Percent Purity 96%
Chemical Name or Material 4,4,4-Trifluorocrotononitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.