missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4,5,5,5-Pentafluoropentanoic acid, Thermo Scientific™
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| CAS | 3637-31-8 |
| Packaging | Amber Glass Bottle |
| Molecular Formula | C5H5F5O2 |
| Quantity | 1 g |
| Synonym | pentanoic acid, 4,4,5,5,5-pentafluoro, pentanoicacid,4,4,5,5,5-pentafluoro |
| InChI Key | HHSIXPRDVJARSI-UHFFFAOYSA-N |
| SMILES | C(CC(C(F)(F)F)(F)F)C(=O)O |
| IUPAC Name | 4,4,5,5,5-pentafluoropentanoic acid |
| Molecular Weight (g/mol) | 192.085 |
| PubChem CID | 23423163 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction