missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-2-[4-(thien-2-ylmethyl)phenyl]-1,3,2-dioxaborolane, 97%, Thermo Scientific™

Product Code. p-3882134
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10707695 250 mg Amber Glass Bottle 250mg
10002084 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10707695 Supplier Thermo Scientific Maybridge Supplier No. CC70639CB
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1007847-76-8
Molecular Formula C17H21BO2S
Molecular Weight (g/mol) 300.22
MDL Number MFCD11841077
InChI Key OPJUSDABOPHYPQ-UHFFFAOYSA-N
Synonym 4,4,5,5-tetramethyl-2-4-thien-2-ylmethyl phenyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-4-thiophen-2-ylmethyl phenyl-1,3,2-dioxaborolane, 4-thien-2-ylmethyl benzeneboronic acid, pinacol ester, 4,4,5,5-tetramethyl-2-4-thiophen-2-yl methyl phenyl-1,3,2-dioxaborolane
PubChem CID 43811052
IUPAC Name 4,4,5,5-tetramethyl-2-[4-(thiophen-2-ylmethyl)phenyl]-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(CC2=CC=CS2)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 300.23
Percent Purity 97%
Chemical Name or Material 4,4,5,5-Tetramethyl-2-[4-(thien-2-ylmethyl)phenyl]-1,3,2-dioxaborolane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.