missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-2-(tetrahydrofuran-3-yl)-1,3,2-dioxaborolane, 97%

Product Code. 11916181 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11916181 1 g Glass bottle 1g
11926181 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11916181 Supplier Thermo Scientific Acros Supplier No. 431100010
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 331958-90-8 | C10H19BO3 | 198.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 331958-90-8
Molecular Formula C10H19BO3
Molecular Weight (g/mol) 198.07
MDL Number MFCD09878900
InChI Key KEBDNHHRQNZXMA-UHFFFAOYNA-N
Synonym 4,4,5,5-tetramethyl-2-tetrahydrofuran-3-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-oxolan-3-yl-1,3,2-dioxaborolane, tetrahydrofuran-3-boronic acid pinacol ester, 4,4,5,5-tetramethyl-2-tetrahydro-3-furanyl-1,3,2-dioxaborolane, tetrahydrofuran-3-ylboronic acid pinacol ester, tetrahydrofuran-3-yl boronic acid pinacol ester, 2-tetrahydrofuran-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl tetrahydrofuran
PubChem CID 11148297
IUPAC Name 4,4,5,5-tetramethyl-2-(oxolan-3-yl)-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1CCOC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 198.07
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4,4,5,5-Tetramethyl-2-(tetrahydrofuran-3-yl)-1,3,2-dioxaborolane
Show More Show Less
compliance-icons
Product Identifier
  • 4,4,5,5-Tetramethyl-2-(tetrahydrofuran-3-yl)-1,3,2-dioxaborolane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.