missing translation for 'onlineSavingsMsg'
Learn More

4-(4,6-Dimethoxy[1.3.5]triazin-2-yl)-4-methylmorpholinium chloride, 97%

Product Code. 10405415 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10405415 25 g Glass bottle 25g
10419802 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10405415 Supplier Thermo Scientific Acros Supplier No. 348960250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3945-69-5 | C10H17ClN4O3 | 276.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3945-69-5
Molecular Formula C10H17ClN4O3
Molecular Weight (g/mol) 276.72
MDL Number MFCD03613550
InChI Key BMTZEAOGFDXDAD-UHFFFAOYSA-M
Synonym dmtmm, 4-4,6-dimethoxy-1,3,5-triazin-2-yl-4-methylmorpholinium chloride, 4-4,6-dimethoxy-1,3,5-triazin-2-yl-4-methyl morpholinium chloride, dmt-mm, 4-4,6-dimethoxy-1,3,5-triazine-2-yl-4-methyl morpholinium chloride, 4-4,6-dimethoxy-1,3,5-triazin-2-yl-4-methylmorpholin-4-ium chloride, kunishima coupling reagent, mmtm, 4-4,6-dimethoxy-1,3,5 triazin-2-yl-4-methyl-morpholin-4-ium chloride
PubChem CID 2734059
SMILES [Cl-].COC1=NC(=NC(OC)=N1)[N+]1(C)CCOCC1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 118°C to 120°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 25 g
IUPAC Name 4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholin-4-ium;chloride
Formula Weight 276.72
Percent Purity 97%
Physical Form Chunks and/or Powder
Chemical Name or Material 4-(4, 6-Dimethoxy[1.3.5]triazin-2-yl)-4-methylmorpholinium chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-(4,6-Dimethoxy[1.3.5]triazin-2-yl)-4-methylmorpholinium chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.