missing translation for 'onlineSavingsMsg'
Learn More

4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride hydrate, 97+%

Product Code. 15435115
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15435115 1 g 1g
15445115 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15435115 Supplier Thermo Scientific Alfa Aesar Supplier No. H26333.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3945-69-5 | C10H17ClN4O3 | 276.72 g/mol

4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride hydrate is used as a pharmaceutical intermediate. It is used as a reagent for activating carboxylic acids in solution and solid phase peptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride hydrate is used as a pharmaceutical intermediate. It is used as a reagent for activating carboxylic acids in solution and solid phase peptide synthesis.

Solubility
Soluble in water, Methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases. Store under dry inert gas. It is sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3945-69-5
Molecular Formula C10H17ClN4O3
Molecular Weight (g/mol) 276.72
MDL Number MFCD03613550
InChI Key BMTZEAOGFDXDAD-UHFFFAOYSA-M
Synonym dmtmm, 4-4,6-dimethoxy-1,3,5-triazin-2-yl-4-methylmorpholinium chloride, 4-4,6-dimethoxy-1,3,5-triazin-2-yl-4-methyl morpholinium chloride, dmt-mm, 4-4,6-dimethoxy-1,3,5-triazine-2-yl-4-methyl morpholinium chloride, 4-4,6-dimethoxy-1,3,5-triazin-2-yl-4-methylmorpholin-4-ium chloride, kunishima coupling reagent, mmtm, 4-4,6-dimethoxy-1,3,5 triazin-2-yl-4-methyl-morpholin-4-ium chloride
PubChem CID 2734059
IUPAC Name 4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholin-4-ium;chloride
SMILES [Cl-].COC1=NC(=NC(OC)=N1)[N+]1(C)CCOCC1

Specifications

Melting Point 110°C to 114°C
Quantity 1 g
UN Number UN3261
Beilstein 8026872
Formula Weight 276.72 (Anhydrous)
Percent Purity ≥97%
Chemical Name or Material 4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride hydrate
compliance-icons compliance-icons
Product Identifier
  • 4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.