missing translation for 'onlineSavingsMsg'
Learn More

4-(4,6-Dimethoxy-s-triazin-2-yl)-4-methyl-morpholinium Chloride, TRC

Product Code. 30388229
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30388229 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388229 Supplier Toronto Research Chemicals Supplier No. TRCD4612455G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(4,6-Dimethoxy-s-triazin-2-yl)-4-methyl-morpholinium Chloride
CAS 3945-69-5
Molecular Formula C10 H17 N4 O3 . Cl
Quantity 5 g
Synonym 4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium Chloride, DMTMM
InChI Formula InChI=1S/C10H17N4O3.ClH/c1-14(4-6-17-7-5-14)8-11-9(15-2)13-10(12-8)16-3;/h4-7H2,1-3H3;1H/q+1;/p-1
SMILES [Cl-].COc1nc(OC)nc(n1)[N+]2(C)CCOCC2
IUPAC Name 4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholin-4-ium;chloride
Molecular Weight (g/mol) 276.72
Formula Weight 276.0989
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.