missing translation for 'onlineSavingsMsg'
Learn More

4-[4-(6-Fluoro-1,2-benzisoxazol-3-yl)piperidin-1-yl]-4-oxobutanoic Acid, TRC

Product Code. 30393138
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30393138 100 mg 100mg
30401151 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30393138 Supplier Toronto Research Chemicals Supplier No. TRCF056450100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[4-(6-Fluoro-1,2-benzisoxazol-3-yl)piperidin-1-yl]-4-oxobutanoic Acid
CAS 1017384-80-3
Molecular Formula C16 H17 F N2 O4
Quantity 100 mg
InChI Formula InChI=1S/C16H17FN2O4/c17-11-1-2-12-13(9-11)23-18-16(12)10-5-7-19(8-6-10)14(20)3-4-15(21)22/h1-2,9-10H,3-8H2,(H,21,22)
SMILES OC(=O)CCC(=O)N1CCC(CC1)c2noc3cc(F)ccc23
IUPAC Name 4-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]-4-oxobutanoic acid
Molecular Weight (g/mol) 320.316
Formula Weight 320.117
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.