missing translation for 'onlineSavingsMsg'
Learn More

4,4,7a-Trimethyl-5,6,7,7a-tetrahydrobenzofuran-2(4H)-one, TRC

Product Code. 30136654
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136654 100 mg 100mg
30134848 50 mg 50mg
30012796 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136654 Supplier Toronto Research Chemicals Supplier No. TRCT896490100MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15356-74-8
Molecular Formula C11 H16 O2
Molecular Weight (g/mol) 180.2435
Synonym 2(4H)-Benzofuranone, 5,6,7,7a-tetrahydro-4,4,7a-trimethyl-, 5,6,7,7a-Tetrahydro-4,4,7a-trimethyl-2(4H)-benzofuranone, (2,6,6-Trimethyl-2-hydroxycyclohexylidene)acetic acid lactone, (±)-Dihydroactinidiolide, 2,4,5,6,7,7a-Hexahydro-4,4,7a-trimethylbenzofuran-2-one, 2-Hydroxy-2,6,6-trimethylcyclohexylidene-1-acetic acid lactone, 4,5,7,7a-Tetrahydro-4,4,7a-trimethyl-2(6H)-benzofuranone, 5,6,7,7α-Tetrahydro-4,4,7α-trimethyl-2(4H)-benzofuranone, 5,8,7,7-Tetrahydro-4,4,7a-trimethyl-α-benzofuranone, 9-Oxa-1,5,5-trimethylbicyclo[4.3.0]non-6-en-8-one, Dihydroactindiolide, Dihydroactinolide
IUPAC Name 4,4,7a-trimethyl-6,7-dihydro-5H-1-benzofuran-2-one
SMILES CC1(C)CCCC2(C)OC(=O)C=C12

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H16O2/c1-10(2)5-4-6-11(3)8(10)7-9(12)13-11/h7H,4-6H2,1-3H3
Formula Weight 180.115
Chemical Name or Material 4,4,7a-Trimethyl-5,6,7,7a-tetrahydrobenzofuran-2(4H)-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.