missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Acetyl-1-piperazinyl)benzeneboronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Product Code. 15435256
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15435256 1 g 1g
15445256 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15435256 Supplier Thermo Scientific Alfa Aesar Supplier No. H59771.03
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1218791-38-8 | Molecular Formula: C18H27BN2O3 | Molecular Weight: 330.235 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1218791-38-8
Molecular Formula C18H27BN2O3
Molecular Weight (g/mol) 330.235
MDL Number MFCD13195756
InChI Key VRZVSHHLWMAHAZ-UHFFFAOYSA-N
Synonym 4-4-acetyl-1-piperazinyl phenylboronic acid pinacol ester, 1-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazin-1-yl ethanone, 4-4-acetylpiperazino phenylboronic acid pinacol ester, 1-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1-piperazinyl ethanone, 4-4-acetyl-1-piperazinyl phenylboronicacidpinacolester, 4-4-acetylpiperazino phenylboronic acid, pinacol ester, 4-4-acetyl-1-piperazinyl benzeneboronic acid pinacol ester, 1-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazin-1-yl ethan-1-one
PubChem CID 53217141
IUPAC Name 1-[4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazin-1-yl]ethanone
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N3CCN(CC3)C(=O)C

Specifications

Quantity 1 g
Formula Weight 330.23
Percent Purity 95%
Chemical Name or Material 4-(4-Acetyl-1-piperazinyl)benzeneboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.