missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Boc-piperazine-1-carbonyl)phenylboronic acid pinacol ester, TRC

Product Code. 30340289
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30340289 100 mg 100mg
30344057 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30340289 Supplier Toronto Research Chemicals Supplier No. TRCB663198100MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(4-Boc-piperazine-1-carbonyl)phenylboronic acid pinacol ester
CAS 864754-13-2
Molecular Formula C22H33BN2O5
Quantity 100 mg
InChI Formula InChI=1S/C22H33BN2O5/c1-20(2,3)28-19(27)25-14-12-24(13-15-25)18(26)16-8-10-17(11-9-16)23-29-21(4,5)22(6,7)30-23/h8-11H,12-15H2,1-7H3
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C(=O)c2ccc(cc2)B3OC(C)(C)C(C)(C)O3
IUPAC Name tert-butyl 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]piperazine-1-carboxylate
Molecular Weight (g/mol) 416.32
Formula Weight 416.2483
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.