missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Bromo-1H-pyrazol-1-yl)aniline, 97%, Thermo Scientific™

Product Code. 10341370
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10341370 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10341370 Supplier Thermo Scientific Maybridge Supplier No. MAY00328DA
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 681441-17-8
Assay Percent Range 97%
Molecular Formula C9H8BrN3
MDL Number MFCD08436686
Quantity 1 g
Synonym 4-4-bromo-1h-pyrazol-1-yl aniline, 4-4-bromopyrazol-1-yl aniline, 4-4-bromopyrazolyl phenylamine
InChI Key HCCGFLDVFPCSOV-UHFFFAOYSA-N
SMILES NC1=CC=C(C=C1)N1C=C(Br)C=N1
IUPAC Name 4-(4-bromopyrazol-1-yl)aniline
Molecular Weight (g/mol) 238.09
PubChem CID 19576898
Formula Weight 238.09
Chemical Name or Material 4-(4-Bromo-1H-pyrazol-1-yl)aniline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.