missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Bromo-1H-pyrazol-1-yl)benzenesulfonyl chloride, 95%, Thermo Scientific™

Product Code. 10775622
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10775622 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10775622 Supplier Thermo Scientific Maybridge Supplier No. MAY00327DA
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1174064-62-0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C9H6BrClN2O2S
Quantity 1 g
Synonym 4-4-bromo-1h-pyrazol-1-yl benzenesulfonyl chloride, 4-4-bromopyrazol-1-yl benzenesulfonyl chloride, 4-4-bromo-1h-pyrazol-1-yl-benzenesulfonyl chloride, 4-4-bromo-1h-pyrazol-1-yl benzene-1-sulfonyl chloride
InChI Key LRYXAJMNOSSJDD-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1N2C=C(C=N2)Br)S(=O)(=O)Cl
IUPAC Name 4-(4-bromopyrazol-1-yl)benzenesulfonyl chloride
Molecular Weight (g/mol) 321.573
PubChem CID 45594324
Formula Weight 321.58
Chemical Name or Material 4-(4-Bromo-1H-pyrazol-1-yl)benzenesulfonyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.