missing translation for 'onlineSavingsMsg'
Learn More

4-[(4-bromothien-2-yl)methyl]morpholine, 97%, Thermo Scientific™

Product Code. 10535944
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10535944 5 g Amber Glass Bottle 5g
10525944 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10535944 Supplier Thermo Scientific Maybridge Supplier No. CC74210DE
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 194851-19-9
Molecular Formula C9H12BrNOS
Molecular Weight (g/mol) 262.17
MDL Number MFCD01653482
InChI Key XYQVGDMXJNORKK-UHFFFAOYSA-N
Synonym 4-4-bromothiophen-2-yl methyl morpholine, 4-4-bromothien-2-yl methyl morpholine, 4-bromo-2-morpholin-4-ylmethyl thiophene, cambridge id 5430345, 4-bromo-2-morpholinomethylthiophene, 4-4-bromo-thiophen-2-ylmethyl-morpholine, morpholine, 4-4-bromo-2-thienyl methyl
PubChem CID 783497
IUPAC Name 4-[(4-bromothiophen-2-yl)methyl]morpholine
SMILES BrC1=CSC(CN2CCOCC2)=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 262.17
Percent Purity 97%
Chemical Name or Material 4-[(4-Bromothien-2-yl)methyl]morpholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.