missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Chlorophenyl)-3-thiosemicarbazide, 97%, Thermo Scientific Chemicals

Product Code. 11363296
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11363296 1 g 1g
11373296 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11363296

Brand: Thermo Scientific Alfa Aesar L12657.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

4-(4-Chlorophenyl)-3-thiosemicarbazide is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(4-Chlorophenyl)-3-thiosemicarbazide is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22814-92-2
Molecular Formula C7H8ClN3S
Molecular Weight (g/mol) 201.672
MDL Number MFCD00041295
InChI Key GFTWJLUVFRTLIL-UHFFFAOYSA-N
Synonym 4-4-chlorophenyl-3-thiosemicarbazide, n-4-chlorophenyl hydrazinecarbothioamide, 3-amino-1-4-chlorophenyl thiourea, 1-amino-3-4-chlorophenyl thiourea, 4-chlorophenyl amino hydrazinomethane-1-thione, acmc-20amw7, maybridge1_006895, 4-p-chlorphenylthiosemicarbazid, 4-4-chlorophenyl thiosemicarbazide, 1-azanyl-3-4-chlorophenyl thiourea
PubChem CID 706988
IUPAC Name 1-amino-3-(4-chlorophenyl)thiourea
SMILES C1=CC(=CC=C1NC(=S)NN)Cl

Specifications

Melting Point ∼180°C
Quantity 1 g
Odor Odorless
UN Number UN2811
Beilstein 1818693
Formula Weight 201.68
Percent Purity 97%
Chemical Name or Material 4-(4-Chlorophenyl)-3-thiosemicarbazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.