missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Chlorophenyl)-4-hydroxypiperidine, TRC

Product Code. 17993165
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17993165 5 g 5g
17902335 100 mg 100mg
17950975 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17993165 Supplier Toronto Research Chemicals Supplier No. TRCC3784805G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39512-49-7
Molecular Formula C11 H14 Cl N O
Molecular Weight (g/mol) 211.69
Synonym 4-(4-Chlorophenyl)piperidin-4-ol, Loperamide Hydrochloride Imp. C (EP)
IUPAC Name 4-(4-chlorophenyl)piperidin-4-ol
SMILES OC1(CCNCC1)c2ccc(Cl)cc2

Specifications

Quantity 5 g
InChI Formula InChI=1S/C11H14ClNO/c12-10-3-1-9(2-4-10)11(14)5-7-13-8-6-11/h1-4,13-14H,5-8H2
Formula Weight 211.0764
Chemical Name or Material 4-(4-Chlorophenyl)piperidin-4-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.