missing translation for 'onlineSavingsMsg'
Learn More

4,4’-Diaminostilbene-2,2’-disulfonic Acid, 95+%, TRC

Product Code. 30361969
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30361969 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30361969 Supplier Toronto Research Chemicals Supplier No. TRCD41683025G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,4'-Diaminostilbene-2,2'-disulfonic Acid
CAS 81-11-8
Molecular Formula C14H14N2O6S2
Quantity 25 g
Synonym 2,2'-(1,2-Ethenediyl)bis[5-aminobenzenesulfonic acid] (ACI), 2,2'-Stilbenedisulfonic acid, 4,4'-diamino- (6CI, 7CI, 8CI), 2,2'-Disulfo-4,4'-stilbenediamine, 4,4'-Diamino-2,2'-stilbenedisulfonic acid, Amsonic acid, DAS, DASD, Diaminostilbenedisulfonic acid, DSD acid, Flavonic acid, NSC 163, p,p'-Diaminodiphenylethylene-o,o'-disulfonic acid, p,p'-Diaminostilbene-o,o'-disulfonic acid
InChI Formula InChI=1S/C14H14N2O6S2/c15-11-5-3-9(13(7-11)23(17,18)19)1-2-10-4-6-12(16)8-14(10)24(20,21)22/h1-8H,15-16H2,(H,17,18,19)(H,20,21,22)/b2-1+
SMILES Nc1ccc(\C=C\c2ccc(N)cc2S(=O)(=O)O)c(c1)S(=O)(=O)O
Molecular Weight (g/mol) 370.4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.