missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dibromobenzil, 97%

Product Code. 11474414
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474414 250 g 250g
11454414 10 g 10g
11464414 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11474414 Supplier Thermo Scientific Alfa Aesar Supplier No. A13090.30
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 35578-47-3 | C14H8Br2O2 | 368.024 g/mol

4,4?-Dibromobenzil was used in preparation of 5,5?-bis -(4-bromo-phenyl)-3-methyl-imidazolidine-2,4-dione, 5,5?- bis -(4-bromo-phenyl)-3-butyl-imidazolidine-2,4-dione, 5,5?-bis-(4-bromo-phenyl)-3-pentyl-imidazolidine-2,4-dione and 5,5?-bis -(4-chloro-phenyl)-3-ethyl imidazolidine-2,4-dione. It was also used in synthesis of novel quinoxaline-based sensitizers for dye-sensitized solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dibromobenzil was used in preparation of 5,5′-bis -(4-bromo-phenyl)-3-methyl-imidazolidine-2,4-dione, 5,5′- bis -(4-bromo-phenyl)-3-butyl-imidazolidine-2,4-dione, 5,5′-bis-(4-bromo-phenyl)-3-pentyl-imidazolidine-2,4-dione and 5,5′-bis -(4-chloro-phenyl)-3-ethyl imidazolidine-2,4-dione. It was also used in synthesis of novel quinoxaline-based sensitizers for dye-sensitized solar cells.

Solubility
Sparingly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35578-47-3
Molecular Formula C14H8Br2O2
MDL Number MFCD00000104
InChI Key NYCBYBDDECLFPE-UHFFFAOYSA-N
PubChem CID 96430

Specifications

Melting Point 224°C to 226°C
Quantity 250 g
Synonym 4,4 '-dibromobenzil, ethanedione, bis 4-bromophenyl, p,p'-dibromobenzil, 1,2-bis 4-bromophenyl ethane-1,2-dione, benzyl, 4,4 '-dibromo, unii-o4txv8a656, bis 4-bromophenyl ethanedione, 1,2-ethanedione, 1,2-bis 4-bromophenyl, bis 4-bromophenyl ethane-1,2-dione, 1,2-bis-4-bromo-phenyl-ethane-1,2-dione
Solubility Information Sparingly soluble aq. soln..
SMILES BrC1=CC=C(C=C1)C(=O)C(=O)C1=CC=C(Br)C=C1
IUPAC Name bis(4-bromophenyl)ethane-1,2-dione
Molecular Weight (g/mol) 368.02
Formula Weight 368.04
Percent Purity 97%
Chemical Name or Material 4,4 '-Dibromobenzil

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.