missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dibromobenzophenone, 98+%

Product Code. 11338035
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11338035 5 g 5g
11328035 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11338035 Supplier Thermo Scientific Alfa Aesar Supplier No. L08200.06
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3988-03-2 | C5H10O2 | 102.13 g/mol

4,4'-Dibromobenzophenone is used in the synthesis of high molecular weight poly(p-phenylene) derivatives through palladium-catalyzed polycondensation reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dibromobenzophenone is used in the synthesis of high molecular weight poly(p-phenylene) derivatives through palladium-catalyzed polycondensation reaction.

Solubility
Soluble in ethanol, 2-propanol, acetone and toluene. Miscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3-2-3988

Specifications

Melting Point 172°C to 176°C
Molecular Formula C5H10O2
MDL Number MFCD00085264
Quantity 5 g
Beilstein 2049958
Synonym ethyl glycidyl ether, 2-ethoxymethyl oxirane, oxirane, ethoxymethyl, 1,2-epoxy-3-ethoxypropane, ethoxymethyl oxirane, 3-ethoxy-1,2-epoxypropane, oxirane, 2-ethoxymethyl, propane, 1,2-epoxy-3-ethoxy, 1-ethoxy-2,3-epoxypropane, ccris 1018
Solubility Information Soluble in ethanol,2-propanol,acetone and toluene. Miscible with water.
InChI Key HQCSZRIVJVOYSU-UHFFFAOYNA-N
SMILES CCOCC1CO1
IUPAC Name 2-(ethoxymethyl)oxirane
Molecular Weight (g/mol) 102.13
PubChem CID 19919
Formula Weight 340.03
Percent Purity ≥98%
Chemical Name or Material 4,4'-Dibromobenzophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.