missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dichlorobutyrophenone, 97%

Product Code. 11490458
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490458 25 g 25g
11309449 50 g 50g
11400468 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490458 Supplier Thermo Scientific Alfa Aesar Supplier No. B24608.14
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 40877-09-6 | C10H10Cl2O | 217.089 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40877-09-6
Molecular Formula C10H10Cl2O
Molecular Weight (g/mol) 217.089
MDL Number MFCD00018993
InChI Key UKCHLVFIVJBCKE-UHFFFAOYSA-N
Synonym 4-chloro-1-4-chlorophenyl butan-1-one, 4,4'-dichlorobutyrophenone, 1-butanone, 4-chloro-1-4-chlorophenyl, gamma,p-dichlorobutyrophenone, 4-chloro-1-4-chlorophenyl-1-oxobutane, 4-chloro-1-4-chlorophenyl-1-butanone, acmc-20a5su, 4',gamma-dichlorobutyrophenone, 4-chloro-4'-chlorobutyrophenone, 4-chloro-4'-chloro-butyrophenone
PubChem CID 2723781
IUPAC Name 4-chloro-1-(4-chlorophenyl)butan-1-one
SMILES C1=CC(=CC=C1C(=O)CCCCl)Cl

Specifications

Melting Point 28°C to 32°C
Refractive Index 1.56
Quantity 25 g
Beilstein 879782
Formula Weight 217.09
Percent Purity 97%
Chemical Name or Material 4,4'-Dichlorobutyrophenone
compliance-icons
Product Identifier
  • 4,4'-Dichlorobutyrophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 4,4'-Dichlorobutyrophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.