missing translation for 'onlineSavingsMsg'
Learn More

4,4-Dimethyl-1-pentene, 99%

Product Code. 10224594 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10224594 25 g Glass bottle 25g
10557454 1 g Glass bottle 1g
10214594 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10224594 Supplier Thermo Scientific Acros Supplier No. 426270250
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 762-62-9 | C7H14 | 98.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 762-62-9
Molecular Formula C7H14
Molecular Weight (g/mol) 98.19
MDL Number MFCD00026337
InChI Key KLCNJIQZXOQYTE-UHFFFAOYSA-N
Synonym 4,4-dimethyl-1-pentene, 1-pentene, 4,4-dimethyl, 2,2-dimethyl-4-pentene, 4,4-dimethyl-pent-1-ene, 4,4-dimethyl pent-1-ene, neoheptaene, acmc-1bcwv, ch3 3cch2ch=ch2, 4,4'-dimethyl-1-pentene
PubChem CID 12984
IUPAC Name 4,4-dimethylpent-1-ene
SMILES CC(C)(C)CC=C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Density 0.6820g/mL
Boiling Point 70°C to 72°C
Flash Point -12°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH2CHCH2C(CH3)3
Quantity 25 g
Specific Gravity 0.682
Formula Weight 98.19
Percent Purity 99%
Chemical Name or Material 4, 4-Dimethyl-1-pentene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4,4-Dimethyl-1-pentene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.